Assay Details
| Gene Symbol: Lrrc75aos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019771.1 | - | 1 - 2 | 173 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 75A | Lrrc75a | Fam211a | Anti-sense | Chr. 11 - 62604884 - 62648583 on NCBI Build GRCm38 | |
| Downstream | membrane magnesium transporter 2 | Mmgt2 | AI852046 | 12.2kb | Sense | Chr. 11 - 62648632 - 62666359 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Lrrc75aos1
Genomic Map