Assay Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028360.2 | NP_082636.3 | 1 - 2 | 222 | 188 | |
| NM_029704.2 | NP_083980.2 | 1 - 2 | 222 | 188 | ||
| GenBank mRNA | AK013368.1 | - | 1 - 2 | 214 | 188 | |
| AK034075.1 | - | 1 - 2 | 212 | 188 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc19
Genomic Map