Assay Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029704.2 | NP_083980.2 | 3 - 4 | 434 | 73 | |
| GenBank mRNA | AK013368.1 | - | 3 - 4 | 426 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc19
Genomic Map