Assay Details
| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028448.1 | NP_082724.1 | 4 - 5 | 692 | 82 | |
| GenBank mRNA | AK014046.1 | - | 4 - 5 | 692 | 82 | |
| BC038868.1 | - | 1 - 2 | 407 | 82 | ||
| BC147794.1 | - | 4 - 5 | 654 | 9087511 | 82 | |
| BC147795.1 | - | 4 - 5 | 654 | 9087512 | 82 | |
| BC147835.1 | - | 4 - 5 | 654 | 9087552 | 82 | |
| BC147837.1 | - | 4 - 5 | 654 | 9087554 | 82 | |
| BP771180.1 | - | 1 - 2 | 82 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
ameboidal-type cell migration
cell cycle
mitotic nuclear division
metabolic process
pericentric heterochromatin assembly
positive regulation of cytokinesis
regulation of chromosome organization
centromere complex assembly
cell division
molecular_function
carbon-sulfur lyase activity
ameboidal-type cell migration
cell cycle
mitotic nuclear division
metabolic process
pericentric heterochromatin assembly
positive regulation of cytokinesis
regulation of chromosome organization
centromere complex assembly
cell division
molecular_function
carbon-sulfur lyase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | BP771180.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | BC038868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | NM_028448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | AK014046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | BP771180.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | BC038868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | NM_028448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P160344 | AK014046.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657294 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915507 | AK138361.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915507 | AK141657.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915507 | BC050989.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915507 | L39770.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915507 | NM_010262.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915507 | Z48800.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391892 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165126 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144108 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131146 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | BP771180.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | BC038868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | NM_028448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | AK014046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | BP771180.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | BC038868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | NM_028448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P160344 | AK014046.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131146 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07367976 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144108 | - | - |
| Whole Mouse Genome G4122A | A_51_P160344 | BP771180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | BC038868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | NM_028448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | AK014046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | BP771180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | BC038868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | NM_028448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P160344 | AK014046.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114082 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144108 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09114013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09557260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09557264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27854448 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10386910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386915 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428706_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386909 | - | - |
| Mouse Genome 430 2.0 Array | 1428706_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428706_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73139_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73139_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100026792.mm.1 | NM_028448.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100026792.mm.1 | NM_028448.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100026792.mm.1 | TC1100002938.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100026793.mm.1 | NM_028448.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100026793.mm.1 | NM_028448.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100026793.mm.1 | TC1100002938.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100026794.mm.1 | NM_028448.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100026794.mm.1 | NM_028448.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100026794.mm.1 | TC1100002938.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428706_at | - | - |
| Mouse Expression Set 430 | 1428706_PM_at | - | - |
| Murine Genome U74v2 Set | 160204_at | - | - |
| Murine Genome U74v2 Set | 160204_PM_at | - | - |
| Murine Genome U74v2 Set | 162491_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162491_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428706_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428706_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428706_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428706_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664729 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664729 | - | - |

Cenpv
Genomic Map