Assay Details
| Gene Symbol: Pigl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039536.2 | NP_001034625.1 | 2 - 3 | 335 | 69 | |
| GenBank mRNA | AK128932.1 | - | 2 - 3 | 335 | 69 | |
| AK158149.1 | - | 2 - 3 | 336 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pigl
Genomic Map