Assay Details
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026842.4 | NP_081118.4 | 6 - 7 | 1299 | 95 | |
| NM_152234.2 | NP_689420.1 | 6 - 7 | 1299 | 95 | ||
| GenBank mRNA | AK048534.1 | - | 6 - 7 | 1220 | 95 | |
| AK049298.1 | - | 6 - 7 | 1054 | 95 | ||
| AK165456.1 | - | 6 - 7 | 1264 | 95 | ||
| AK172695.1 | - | 6 - 7 | 1268 | 95 | ||
| BC010213.1 | - | 3 - 4 | 642 | 95 | ||
| BC026847.1 | - | 6 - 7 | 1282 | 4015641 | 95 | |
| BC028857.1 | - | 6 - 7 | 1238 | 4507759 | 95 | |
| BC051098.1 | - | 5 - 6 | 927 | 95 | ||
| BC080667.1 | - | 6 - 7 | 1260 | 4935292 | 95 | |
More Information
Set Membership: |
|

Ubqln1
Genomic Map