Assay Details
| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173030.2 | NP_766618.2 | 11 - 12 | 1905 | 99 | |
| GenBank mRNA | AB082928.1 | - | 9 - 10 | 1393 | 99 | |
| AK045417.1 | - | 11 - 12 | 1906 | 99 | ||
| BC131651.1 | - | 11 - 12 | 1584 | 40110517 | 99 | |
| BC131652.1 | - | 10 - 11 | 1615 | 40110518 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt13
Genomic Map