Assay Details
| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173030.2 | NP_766618.2 | 2 - 3 | 415 | 123 | |
| GenBank mRNA | AK038387.1 | - | 2 - 3 | 416 | 123 | |
| AK045417.1 | - | 2 - 3 | 416 | 123 | ||
| BC131651.1 | - | 2 - 3 | 94 | 40110517 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038387.1, AK045417.1, BC131651.1, NM_173030.2 transcripts.
Set Membership: |
|

Galnt13
Genomic Map