Assay Details
| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173030.2 | NP_766618.2 | 3 - 4 | 658 | 80 | |
| GenBank mRNA | AB082928.1 | - | 1 - 2 | 146 | 80 | |
| AK030726.1 | - | 2 - 3 | 588 | 80 | ||
| AK038387.1 | - | 3 - 4 | 659 | 80 | ||
| AK045417.1 | - | 3 - 4 | 659 | 80 | ||
| BC131651.1 | - | 3 - 4 | 337 | 40110517 | 80 | |
| BC131652.1 | - | 2 - 3 | 368 | 40110518 | 80 | |
More Information
Set Membership: |
|

Galnt13
Genomic Map