Assay Details
| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173030.2 | NP_766618.2 | 6 - 7 | 1193 | 92 | |
| GenBank mRNA | AB082928.1 | - | 4 - 5 | 681 | 92 | |
| AK030726.1 | - | 5 - 6 | 1125 | 92 | ||
| AK038387.1 | - | 6 - 7 | 1194 | 92 | ||
| AK045417.1 | - | 6 - 7 | 1194 | 92 | ||
| BC131651.1 | - | 6 - 7 | 872 | 40110517 | 92 | |
| BC131652.1 | - | 5 - 6 | 903 | 40110518 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt13
Genomic Map