| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200034652.mm.1 |
NM_001166597.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034652.mm.1 |
NM_001166597.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034652.mm.1 |
TC0200004130.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034649.mm.1 |
NM_001166597.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034649.mm.1 |
NM_001166597.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034649.mm.1 |
TC0200004130.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034648.mm.1 |
NM_001166597.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034648.mm.1 |
NM_001166597.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034648.mm.1 |
TC0200004130.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034651.mm.1 |
NM_001166597.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034651.mm.1 |
NM_001166597.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034651.mm.1 |
TC0200004130.mm.1 |
MTA TC |