Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 2 - 3 | 878 | 78 | |
| GenBank mRNA | AK087503.1 | - | 2 - 3 | 879 | 78 | |
| BC016217.1 | - | 1 - 2 | 1224 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map