Assay Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110981.1 | 3 - 4 | 405 | 74 | ||
| GenBank mRNA | AK003191.1 | - | 1 - 2 | 104 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003191.1 transcripts.
Set Membership: |
|

Slc39a13
Genomic Map