Assay Details
| Gene Symbol: Gpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081089.2 | NP_001074558.2 | 7 - 8 | 678 | 79 | |
| GenBank mRNA | AB257502.1 | - | 6 - 7 | 596 | 79 | |
| AK131154.1 | - | 7 - 8 | 677 | 79 | ||
| BC113165.1 | - | 7 - 8 | 672 | 40059976 | 79 | |
| BC113776.1 | - | 7 - 8 | 672 | 40059969 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpat2
Genomic Map