Assay Details
| Gene Symbol: Siglech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080658.1 | - | 1 - 2 | 537 | 79 | |
| BC144741.1 | - | 1 - 2 | 485 | 9053289 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglech
Genomic Map