Assay Details
Gene Symbol: 2010107E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027360.2 | NP_081636.1 | 2 - 3 | 175 | 126 | |
GenBank mRNA | AK008333.1 | - | 2 - 3 | 176 | 126 | |
BC055712.1 | - | 2 - 3 | 154 | 6408438 | 126 | |
BC059719.1 | - | 2 - 3 | 163 | 6774233 | 126 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
