Assay Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146073.3 | NP_666185.3 | 3 - 4 | 1407 | 56 | |
| GenBank mRNA | AF542387.1 | - | 4 - 5 | 1076 | 56 | |
| AK041496.1 | - | 3 - 4 | 1090 | 56 | ||
| AK041648.1 | - | 4 - 5 | 1403 | 56 | ||
| AK046719.1 | - | 3 - 4 | 1395 | 56 | ||
| AK080845.1 | - | 3 - 4 | 1411 | 56 | ||
| BC059814.1 | - | 3 - 4 | 1450 | 6813525 | 56 | |
| BC086758.1 | - | 4 - 5 | 463 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc14
Genomic Map