Assay Details
| Gene Symbol: Tdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028354.4 | NP_082630.2 | 12 - 13 | 1484 | 60 | |
| GenBank mRNA | AK013427.1 | - | 11 - 12 | 959 | 60 | |
| AK014855.1 | - | 12 - 13 | 1464 | 60 | ||
| AK033868.1 | - | 12 - 13 | 1482 | 60 | ||
| BC019804.1 | - | 9 - 10 | 808 | 4018534 | 60 | |
| BC065162.1 | - | 11 - 12 | 1454 | 6402921 | 60 | |
More Information
Set Membership: |
|

Tdp1
Genomic Map