Assay Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011246.2 | NP_035376.1 | 1 - 2 | 206 | 77 | |
| GenBank mRNA | AF106070.1 | - | 1 - 2 | 50 | 77 | |
| AK013548.1 | - | 1 - 2 | 252 | 77 | ||
| AK141524.1 | - | 1 - 2 | 331 | 77 | ||
| BC057120.1 | - | 1 - 2 | 206 | 6831137 | 77 | |
| BC057341.1 | - | 1 - 2 | 206 | 6831137 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp1
Genomic Map