Assay Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011246.2 | NP_035376.1 | 3 - 4 | 497 | 97 | |
| GenBank mRNA | AF106070.1 | - | 3 - 4 | 341 | 97 | |
| AK013548.1 | - | 3 - 4 | 543 | 97 | ||
| AK134005.1 | - | 3 - 4 | 511 | 97 | ||
| AK141524.1 | - | 3 - 4 | 622 | 97 | ||
| BC057120.1 | - | 3 - 4 | 497 | 6831137 | 97 | |
| BC057341.1 | - | 3 - 4 | 497 | 6831137 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rasgrp1
Genomic Map