Assay Details
Gene Symbol: 3200001D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045888.2 | 1 - 2 | 470 | 69 | ||
GenBank mRNA | AK014278.1 | - | 1 - 2 | 471 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | eukaryotic translation initiation factor 1A pseudogene | Gm5039 | EG266459 | -46.8kb | Sense | Chr. 12 - 88320122 - 88321762 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930473H19 gene | 4930473H19Rik | 53.9kb | Sense | Chr. 12 - 88431192 - 88444976 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
