Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1431850_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431850_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431850_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1200014863.mm.1 |
NR_045888.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014863.mm.1 |
TC1200002141.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1431850_at |
- |
- |
Mouse Expression Set 430 |
1431850_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1431850_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1431850_at |
- |
- |