Assay Details
| Gene Symbol: Gm29508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076979.1 | - | 2 - 3 | 481 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 181, member A |
Fam181a | EG544888 | Anti-sense | Chr. 12 - 103311013 - 103317067 on NCBI Build GRCm38 | |
| Downstream | ankyrin repeat and SOCS box-containing 2 | Asb2 | 1110008E15Rik | 8.2kb | Sense | Chr. 12 - 103321142 - 103356001 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm29508
Genomic Map