Assay Details
| Gene Symbol: Ighg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037822.1 | - | 1374 | 66 | ||
| AK131185.1 | - | 1561 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ighg3| Gene Symbol: Ighg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037822.1 | - | 1374 | 66 | ||
| AK131185.1 | - | 1561 | 66 | |||
Set Membership: |
|