Assay Details
| Gene Symbol: A530006G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046014.1 | 1 - 2 | 180 | 97 | ||
| GenBank mRNA | AK079921.1 | - | 1 - 2 | 181 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired box 1 | Pax1 |
Pax-1, hbs, hunchback, un, undulated, wt |
-335.6kb | Sense | Chr. 2 - 147361925 - 147375049 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9030622O22 gene | 9030622O22Rik | 233.4kb | Anti-sense | Chr. 2 - 147944459 - 148040560 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A530006G24Rik
Genomic Map