Assay Details
| Gene Symbol: 9030622O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018565.1 | - | 3 - 4 | 761 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A530006G24 gene | A530006G24Rik | Gm1003 | -245.0kb | Anti-sense | Chr. 2 - 147710693 - 147717632 on NCBI Build GRCm38 |
| Downstream | forkhead box A2 | Foxa2 |
HNF3-beta, HNF3beta, Hnf-3b, Hnf3b, Tcf-3b, Tcf3b |
75.6kb | Sense | Chr. 2 - 148042877 - 148046969 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

9030622O22Rik
Genomic Map