Assay Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024618.2 | NP_001019789.1 | 2 - 3 | 307 | 56 | |
| NM_001083919.1 | NP_001077388.1 | 2 - 3 | 307 | 56 | ||
| GenBank mRNA | AK040786.1 | - | 2 - 3 | 308 | 56 | |
| AY775570.1 | - | 1 - 2 | 161 | 56 | ||
| DQ011666.1 | - | 2 - 3 | 308 | 56 | ||
| EF119712.1 | - | 2 - 3 | 307 | 56 | ||
| EF119713.1 | - | 463 | 56 | |||
| EF119714.1 | - | 511 | 56 | |||
| EF119715.1 | - | 538 | 56 | |||
| EF119716.1 | - | 2 - 3 | 307 | 56 | ||
| EF122140.1 | - | 1 - 2 | 62 | 56 | ||
| EU286531.1 | - | 2 - 3 | 290 | 56 | ||
| KM273012.1 | - | 749 | 56 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Xirp2
Genomic Map