Assay Details
Gene Symbol: D330050G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040335.1 | 2 - 3 | 280 | 100 | ||
GenBank mRNA | AK083230.1 | - | 2 - 3 | 258 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane and coiled-coil domains 5 | Tmco5 |
1700095F04Rik, Tmco5a |
-8.5kb | Sense | Chr. 2 - 116878691 - 116892500 on NCBI Build GRCm38 |
Downstream |
sprouty protein with EVH-1 domain 1, related sequence |
Spred1 |
5730461F13Rik, AL024345, AW047647 |
210.2kb | Sense | Chr. 2 - 117121071 - 117182279 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
