Assay Details
| Gene Symbol: Hrnr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133698.2 | NP_598459.2 | 1 - 2 | 56 | 86 | |
| GenBank mRNA | AK041026.1 | - | 1 - 2 | 57 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041026.1, NM_133698.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963094 | BC118008.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963094 | BC118008.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963094 | BC118008.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963094 | AY027660.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963094 | AY027660.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | AF508157.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | AK138669.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | AK154161.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | AK168226.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | BC003903.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | BC057964.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | BC062201.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610478 | NM_133994.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147644 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963094 | BC118008.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963094 | BC118008.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963094 | BC118008.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963094 | AY027660.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963094 | AY027660.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482781 | AK004070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482781 | AY027660.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482781 | NM_133698.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248986 | AY027660.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248986 | NM_133698.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108376 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493965 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493978 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451613_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493925 | - | - |
| Mouse Genome 430 2.0 Array | 1451613_at | - | - |
| Mouse Genome 430 2.0 Array | 1451613_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68723_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68723_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006510.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006510.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006510.mm.1 | TC0300000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006514.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006514.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006514.mm.1 | TC0300000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006506.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006506.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006506.mm.1 | TC0300000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006508.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006508.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006508.mm.1 | TC0300000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006511.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006511.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006511.mm.1 | TC0300000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006509.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006509.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006509.mm.1 | TC0300000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006513.mm.1 | NM_133698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006513.mm.1 | NM_133698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006513.mm.1 | TC0300000846.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451613_at | - | - |
| Mouse Expression Set 430 | 1451613_PM_at | - | - |
| Murine Genome U74v2 Set | 166991_PM_at | - | - |
| Murine Genome U74v2 Set | 166991_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451613_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451613_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 733573 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 780121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2603101 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2603101 | - | - |
| mouse-6 v1.1 (Illumina) | scl22915.1.3052_68-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22915.1.3052_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2603101 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2603101 | - | - |

Hrnr
Genomic Map