Assay Details
Gene Symbol: LOC102639987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK045137.1 | - | 4 - 5 | 619 | 136 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FERM domain containing 4A | Frmd4a |
2700017I06Rik, 5330420J21, AI594879, C230040M21Rik, Frmd4, Gm13190, OTTMUSG00000010806, mKIAA1294 |
Anti-sense | Chr. 2 - 4017695 - 4614043 on NCBI Build GRCm38 | |
Downstream | predicted gene 13179 | Gm13179 | OTTMUSG00000010754 | 294.1kb | Sense | Chr. 2 - 4586449 - 4594529 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
