Assay Details
| Gene Symbol: Dbhos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040524.1 | 2 - 3 | 768 | 170 | ||
| GenBank mRNA | AK032445.1 | - | 2 - 3 | 770 | 170 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 163, member B |
Fam163b | C630035N08Rik | -18.8kb | Sense | Chr. 2 - 27110379 - 27142477 on NCBI Build GRCm38 |
| Downstream | dopamine beta hydroxylase | Dbh | 3.7kb | Anti-sense | Chr. 2 - 27165507 - 27183204 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300002762.rn
Set Membership: |
|

Dbhos
Genomic Map