Assay Details
| Gene Symbol: F730016J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036396.1 | - | 1 - 2 | 60 | 68 | |
| AK089376.1 | - | 1 - 2 | 54 | 68 | ||
| AK089406.1 | - | 1 - 2 | 54 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ficolin B | Fcnb | Fcn2 | -10.6kb | Anti-sense | Chr. 2 - 28076479 - 28084913 on NCBI Build GRCm38 |
| Downstream | olfactomedin 1 | Olfm1 |
AMY, AW742568, Noe1, OlfA, Pancortin, Pancortin3 |
93.1kb | Sense | Chr. 2 - 28193093 - 28230736 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

F730016J06Rik
Genomic Map