Assay Details
| Gene Symbol: Ttc7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033213.1 | NP_001028385.1 | 10 - 11 | 1371 | 66 | |
| GenBank mRNA | AK046740.1 | - | 10 - 11 | 1341 | 66 | |
More Information
Set Membership: |
|

Ttc7b
Genomic Map