Assay Details
| Gene Symbol: Ttc7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033213.1 | NP_001028385.1 | 16 - 17 | 2001 | 106 | |
| GenBank mRNA | AK046740.1 | - | 16 - 17 | 1971 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc7b
Genomic Map