Assay Details
| Gene Symbol: Ttc7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033213.1 | NP_001028385.1 | 4 - 5 | 698 | 76 | |
| GenBank mRNA | AK046740.1 | - | 4 - 5 | 667 | 76 | |
| AK148363.1 | - | 4 - 5 | 670 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc7b
Genomic Map