Assay Details
| Gene Symbol: Gm13580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046065.1 | 1 - 2 | 196 | 76 | ||
| GenBank mRNA | BC051520.1 | - | 1 - 2 | 205 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lymphocyte antigen 75 | Ly75 |
CD205, DEC-205, DEC205 |
-28.3kb | Anti-sense | Chr. 2 - 60293760 - 60383231 on NCBI Build GRCm38 |
| Downstream | phospholipase A2 receptor 1 | Pla2r1 |
Pla2-r, Pla2g1br, Pla2r |
5.5kb | Anti-sense | Chr. 2 - 60417543 - 60553350 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm13580
Genomic Map