Assay Details
| Gene Symbol: 1700048B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006721.1 | - | 2 - 3 | 490 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930518P08 gene | 4930518P08Rik | -148.7kb | Sense | Chr. 13 - 50753393 - 50765248 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 9430083A17 gene | 9430083A17Rik | 173.0kb | Sense | Chr. 13 - 51097738 - 51100727 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

1700048B10Rik
Genomic Map