Assay Details
| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036449.1 | 1 - 2 | 938 | 62 | ||
| GenBank mRNA | AK037190.1 | - | 1 - 2 | 939 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037190.1 transcripts.
Set Membership: |
|

Gm5084
Genomic Map