Assay Details
| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036449.1 | 2 - 3 | 1509 | 69 | ||
| GenBank mRNA | AK037190.1 | - | 2 - 3 | 1510 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037190.1 transcripts.
Set Membership: |
|

Gm5084
Genomic Map