Assay Details
| Gene Symbol: Entpd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028093.1 | NP_082369.1 | 9 - 10 | 1546 | 61 | |
| GenBank mRNA | AK008590.1 | - | 5 - 6 | 1068 | 61 | |
| AY364442.1 | - | 9 - 10 | 1408 | 61 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989215 | NM_028093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989215 | AV026718.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926089 | AF309644.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926089 | AK003118.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926089 | BC057975.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926089 | NM_008993.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480354 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180571 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989215 | NM_028093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989215 | AV026718.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P468482 | BC031143.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P468482 | NM_028093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P468482 | AK008590.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P468482 | AV026718.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P468482 | AY364442.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180571 | - | - |
| Whole Mouse Genome G4122A | A_51_P468482 | BC031143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P468482 | NM_028093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P468482 | AK008590.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P468482 | AV026718.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P468482 | AY364442.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180571 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01314902 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01314892 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01314889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01887213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01887211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01887218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01225177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01166868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01979238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01166861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01879188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01314902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01166838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01979239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01887210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01166869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069683 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10469930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469931 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429550_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480585 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10469923 | - | - |
| Mouse Genome 430 2.0 Array | 1429550_at | - | - |
| Mouse Genome 430 2.0 Array | 1429550_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72090_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72090_at | - | - |
| Mouse Expression Set 430 | 1429550_PM_at | - | - |
| Mouse Expression Set 430 | 1429550_at | - | - |
| Murine Genome U74v2 Set | 165683_PM_at | - | - |
| Murine Genome U74v2 Set | 165683_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429550_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429550_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 407466 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4920079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2744998 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2744998 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2744998 | - | - |

Entpd8
Genomic Map