Assay Details
| Gene Symbol: Arhgap44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099288.1 | NP_001092758.1 | 5 - 6 | 717 | 71 | |
| NM_175003.3 | NP_778168.2 | 5 - 6 | 717 | 71 | ||
| GenBank mRNA | AK053220.1 | - | 5 - 6 | 445 | 71 | |
| BC056366.1 | - | 5 - 6 | 717 | 6825221 | 71 | |
| BC059911.1 | - | 5 - 6 | 717 | 6825221 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap44
Genomic Map