Assay Details
| Gene Symbol: Arhgap44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099288.1 | NP_001092758.1 | 15 - 16 | 1662 | 65 | |
| NM_175003.3 | NP_778168.2 | 15 - 16 | 1662 | 65 | ||
| GenBank mRNA | AK079081.1 | - | 10 - 11 | 963 | 65 | |
| BC056366.1 | - | 15 - 16 | 1662 | 6825221 | 65 | |
| BC059911.1 | - | 15 - 16 | 1662 | 6825221 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap44
Genomic Map