Assay Details
| Gene Symbol: Stkld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198628.2 | NP_941030.2 | 18 - 19 | 1932 | 90 | |
| GenBank mRNA | BC050806.1 | - | 18 - 19 | 1933 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994633 | BC050806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994633 | NM_198628.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38758 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145839 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130610 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994633 | BC050806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994633 | NM_198628.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081283 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04461626 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130610 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04461657 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145839 | - | - |
| Whole Mouse Genome G4122A | A_52_P415267 | BC050806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P415267 | NM_198628.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137397 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145839 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01866910 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01866922 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01866916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01762672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01278956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01866926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01278958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01637482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02264296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081279 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10470334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470337 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470344 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470339 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470335 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470341 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470329 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200003049.mm.1 | NM_198628.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003049.mm.1 | NM_198628.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200003049.mm.1 | TC0200000460.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 166105_PM_at | - | - |
| Murine Genome U74v2 Set | 166105_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 377750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3190746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190746 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641925 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2641925 | - | - |
| mouse-6 v1.1 (Illumina) | scl0279029.18_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190746 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0279029.18_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2641920 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641920 | - | - |

Stkld1
Genomic Map