Assay Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114088.2 | NP_001107560.1 | 7 - 8 | 663 | 99 | |
| GenBank mRNA | AK010339.1 | - | 5 - 6 | 99 | ||
| AK033091.1 | - | 655 | 99 | |||
| AK142933.1 | - | 7 - 8 | 647 | 99 | ||
| AK167476.1 | - | 7 - 8 | 655 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdlim7
Genomic Map