Assay Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084820.1 | - | 1 - 2 | 138 | 100 | |
| AK160104.1 | - | 1 - 2 | 108 | 100 | ||
| AK170084.1 | - | 1 - 2 | 119 | 100 | ||
More Information
Set Membership: |
|

Tulp4
Genomic Map