Assay Details
| Gene Symbol: Grin1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242940.1 | NP_001229869.1 | 3 - 4 | 544 | 65 | |
| GenBank mRNA | AK087384.1 | - | 3 - 4 | 544 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 26 | Lrrc26 | -4.7kb | Sense | Chr. 2 - 25289911 - 25291193 on NCBI Build GRCm38 | |
| Downstream | expressed sequence AA543186 | AA543186 | 28.5kb | Anti-sense | Chr. 2 - 25327450 - 25332571 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001242940.1 transcripts.
Set Membership: |
|

Grin1os
Genomic Map