Assay Details
| Gene Symbol: 5430427M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045858.1 | 3 - 4 | 284 | 77 | ||
| GenBank mRNA | AK017349.1 | - | 3 - 4 | 285 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26512 |
Gm26512 | -240.6kb | Sense | Chr. 12 - 90738212 - 90812245 on NCBI Build GRCm38 | |
| Downstream | thyroid stimulating hormone receptor | Tshr |
AI481368, hypothroid, hyt, pet |
344.4kb | Sense | Chr. 12 - 91400993 - 91540509 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

5430427M07Rik
Genomic Map