Assay Details
| Gene Symbol: Mirg | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028265.1 | 5 - 6 | 591 | 62 | |||
| GenBank mRNA | AJ517767.1 | - | LIT1749 Mirg |
5 - 6 | 591 | 62 | |
| EU434920.1 | - | 2476 | 62 | ||||
| EU616813.1 | - | 3088 | 62 | ||||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 369 | Mir369 |
Mirn369, mir-369, mmu-mir-369 |
Sense | Chr. 12 - 109743418 - 109743496 on NCBI Build GRCm38 | |
| Downstream | microRNA 3072 | Mir3072 |
mir-3072, mmu-mir-3072 |
4.0kb | Sense | Chr. 12 - 109747878 - 109747960 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mirg
Genomic Map