Assay Details
| Gene Symbol: Mirg | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028265.1 | 6 - 7 | 770 | 66 | |||
| GenBank mRNA | AF498297.1 | - | 1 - 2 | 42 | 66 | ||
| AF498298.1 | - | 1 - 2 | 42 | 66 | |||
| AJ517767.1 | - | LIT1749 Mirg |
6 - 7 | 770 | 66 | ||
| AK013406.1 | - | 1 - 2 | 67 | 66 | |||
| EU434920.1 | - | 2655 | 66 | ||||
| EU616812.1 | - | 3292 | 66 | ||||
| EU616813.1 | - | 3267 | 66 | ||||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3072 | Mir3072 |
mir-3072, mmu-mir-3072 |
Sense | Chr. 12 - 109747878 - 109747960 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 3110009F21 gene | 3110009F21Rik | 403.4kb | Anti-sense | Chr. 12 - 110151654 - 110158073 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Mirg
Genomic Map