Assay Details
| Gene Symbol: Sp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018042.3 | NP_001018052.1 | 3 - 4 | 476 | 144 | |
| NM_001098425.1 | NP_001091895.1 | 1 - 2 | 385 | 144 | ||
| GenBank mRNA | AK088238.1 | - | 1 - 2 | 360 | 144 | |
| AK132702.1 | - | 1 - 2 | 131 | 144 | ||
| BC079874.1 | - | 3 - 4 | 476 | 6833706 | 144 | |
| CA328978.1 | - | 1 - 2 | 389 | 144 | ||
| CJ167744.1 | - | 1 - 2 | 386 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132702.1, NM_001098425.1 transcripts.
Set Membership: |
|

Sp3
Genomic Map